Please login to your online account to display your discounted pricing

2-N-Methyl-2-pyridylaminoethanol, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-N-Methyl-2-pyridylaminoethanol
CAS 122321-04-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8 H12 N2 O
Formula Weight 152.2
MDL Number MFCD02667633
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC373150050

  SDS
fisher scientific acros
AC373150050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC373150250

  SDS
thermo fisher scientific
AC373150250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 152.2
Physical Form Viscous Liquid
Quantity 5g
Color Clear light yellow to light brown
CAS Max % 100.0
Refractive Index 1.5705 to 1.5735 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Molecular Formula C8 H12 N2 O
CAS 122321-04-4
Assay Percent Range 98%
CAS Min % 97.5
Packaging Glass bottle
Chemical Name or Material 2-N-Methyl-2-pyridylaminoethanol
MDL Number MFCD02667633
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.