Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Hexylpyridine, 98%

$810.64 - $810.64

Chemical Identifiers

Chemical Name or Material 2-n-Hexylpyridine
CAS 1129-69-7
Molecular Formula C11H17N
Formula Weight 163.27
MDL Number MFCD00023552
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1277214

  SDS
alfa aesar™
A1277214
25g Each for $810.64
Description & Specifications

Specifications

Formula Weight 163.27
Chemical Name or Material 2-n-Hexylpyridine
Assay 98%
Molecular Formula C11H17N
MDL Number MFCD00023552
CAS 1129-69-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UT4245000
Quantity 25g

Applications
2-n-Hexylpyridine is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Slightly soluble in water.

Notes
This product is relativelyunstable under normal temperature. Keep away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.