Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Hexylcyclopentanone, 97%

$37.41 - $137.14

Chemical Identifiers

Chemical Name or Material 2-n-Hexylcyclopentanone
CAS 13074-65-2
Molecular Formula C11H20O
Formula Weight 168.28
MDL Number MFCD00059516
Beilstein 2041950
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1011206

  SDS
alfa aesar™
L1011206
5g Each for $37.41

AAL1011214

  SDS
alfa aesar™
L1011214
25g Each for $137.14
Description & Specifications

Specifications

Formula Weight 168.28
Chemical Name or Material 2-n-Hexylcyclopentanone
TSCA TSCA
Assay 97%
Molecular Formula C11H20O
MDL Number MFCD00059516
Beilstein 2041950
Flash Point 112°C (233°F)
Boiling Point 108° to 110°C (10mmHg)
Quantity 5g
CAS 13074-65-2
RTECS Number GY4972000