Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Butylpropane-1,3-diol, 97%

$209.34 - $209.34

Chemical Identifiers

Chemical Name or Material 2-n-Butylpropane-1,3-diol
CAS 2612-26-2
Molecular Formula CH3(CH2)3CH(CH2OH)2
Formula Weight 132.2
MDL Number MFCD00799395
Synonym 2-(Hydroxymethyl)hexan-1-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2173906

 
alfa aesar™
B2173906
5g Each for $209.34
Description & Specifications

Specifications

Formula Weight 132.2
MDL Number MFCD00799395
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Chemical Name or Material 2-n-Butylpropane-1,3-diol
Assay 97%
Molecular Formula CH3(CH2)3CH(CH2OH)2
CAS 2612-26-2
Synonym 2-(Hydroxymethyl)hexan-1-ol
Health Hazard 1 Danger
Quantity 5g
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.