Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Butylbenzo[b]furan, 98%

$34.32 - $105.89

Chemical Identifiers

Chemical Name or Material 2-n-Butylbenzo[b]furan
CAS 4265-27-4
Molecular Formula C12H14O
Formula Weight 174.24
MDL Number MFCD00005851
Beilstein 1365111
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0780506

  SDS
alfa aesar™
L0780506
5g Each for $34.32

AAL0780514

  SDS
alfa aesar™
L0780514
25g Each for $105.89
Description & Specifications

Specifications

Formula Weight 174.24
Chemical Name or Material 2-n-Butylbenzo[b]furan
Molecular Formula C12H14O
Assay 98%
MDL Number MFCD00005851
CAS 4265-27-4
Boiling Point 114° to 116°C (8mmHg)
Beilstein 1365111
Quantity 5g
Flash Point 101°C (213°F)