Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Butyl-5-methylthiophene, 97%

$29.86 - $369.43

Chemical Identifiers

Chemical Name or Material 2-n-Butyl-5-methylthiophene
CAS 111510-96-4
Molecular Formula C9H14S
Formula Weight 154.28
MDL Number MFCD00091566
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2494303

  SDS
alfa aesar™
B2494303
1g Each for $29.86

AAB2494306

  SDS
alfa aesar™
B2494306
5g Each for $101.28

AAB2494314

  SDS
alfa aesar™
B2494314
25g Each for $369.43
Description & Specifications

Specifications

Formula Weight 154.28
Chemical Name or Material 2-n-Butyl-5-methylthiophene
Assay 97%
Molecular Formula C9H14S
MDL Number MFCD00091566
CAS 111510-96-4
Quantity 1g