Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Butyl-5-ethylthiophene, 97%

$68.70 - $68.70

Chemical Identifiers

Chemical Name or Material 2-n-Butyl-5-ethylthiophene
CAS 54411-06-2
Molecular Formula C10H16S
Formula Weight 168.3
MDL Number MFCD00053118
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2270806

  SDS
alfa aesar™
B2270806
5g Each for $68.70
Description & Specifications

Specifications

Formula Weight 168.3
Chemical Name or Material 2-n-Butyl-5-ethylthiophene
Assay 97%
Molecular Formula C10H16S
MDL Number MFCD00053118
CAS 54411-06-2
Quantity 5g