Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Butoxypyridine, 95%

$61.64 - $188.35

Chemical Identifiers

Chemical Name or Material 2-n-Butoxypyridine
CAS 27361-16-6
Molecular Formula C9H13NO
Formula Weight 151.21
MDL Number MFCD00006267
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2165114

  SDS
alfa aesar™
B2165114
25g Each for $61.64

AAB2165122

  SDS
alfa aesar™
B2165122
100g Each for $188.35
Description & Specifications

Specifications

Formula Weight 151.21
MDL Number MFCD00006267
Chemical Name or Material 2-n-Butoxypyridine
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25g
CAS 27361-16-6
Health Hazard 1 Warning
Assay 95%
Boiling Point 199° to 201°C
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Molecular Formula C9H13NO
Flash Point 75°C (167°F)
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.