Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(N-Boc-methylamino)-5-iodo-3-methylpyridine, 95%

$186.84 - $186.84

Chemical Identifiers

Chemical Name or Material 2-(N-Boc-methylamino)-5-iodo-3-methylpyridine
CAS 942206-08-8
Molecular Formula C12H17IN2O2
Formula Weight 348.18
MDL Number MFCD09037457
Synonym tert-Butyl N-methyl-N-(5-iodo-3-methyl-2-pyridyl)carbamate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27397MD

  SDS
alfa aesar™
H27397MD
250mg Each for $186.84
Description & Specifications

Specifications

Formula Weight 348.18
MDL Number MFCD09037457
Synonym tert-Butyl N-methyl-N-(5-iodo-3-methyl-2-pyridyl)carbamate
Chemical Name or Material 2-(N-Boc-methylamino)-5-iodo-3-methylpyridine
Quantity 250mg
Melting Point 97° to 100°C
Molecular Formula C12H17IN2O2
CAS 942206-08-8
Assay 95%