Please login to your online account to display your discounted pricing

2-MPMDQ, Tocris

Potent, selective α1 antagonist; Also σ2 ligand

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material 2-MPMDQ
Assay Percent Range >98%
Molecular Formula C23H27N5O2
Formula Weight 405.5
Synonym 2-[[4-(2-Methoxyphenyl)piperazin-1-yl]methyl-6-methyl-2, 3-dihydroimidazo[1, 2c]quinazolin-5(6H)-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

066110

 
r & d systems
066110
10mg Each for $181.19

066150

 
r & d systems
066150
50mg Each for $762.58
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 405.5
Chemical Name or Material 2-MPMDQ
Assay Percent Range >98%
Molecular Formula C23H27N5O2
CAS 149847-77-8
Synonym 2-[[4-(2-Methoxyphenyl)piperazin-1-yl]methyl-6-methyl-2, 3-dihydroimidazo[1, 2c]quinazolin-5(6H)-one
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature