missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Morpholinothio)benzothiazole 97.0+%, TCI America™

Manufacturer:  TCI America M0532500G

 View more versions of this product

Catalog No. M0532500G



Specifications

Specifications

2-(Morpholinothio)benzothiazole
86°C
102-77-2
MFCD00022870
N-Oxydiethylene-2-benzothiazolesulfenamide
C1COCCN1SC2=NC3=CC=CC=C3S2
252.35
252.35
≥97.0% (GC)
White-Yellow
500g
C11H12N2OS2
3077
MHKLKWCYGIBEQF-UHFFFAOYSA-N
4-(1,3-benzothiazol-2-ylsulfanyl)morpholine
7619
Crystal-Powder at 20°C

Chemical Identifiers

102-77-2
252.35
MHKLKWCYGIBEQF-UHFFFAOYSA-N
7619
C1COCCN1SC2=NC3=CC=CC=C3S2
C11H12N2OS2
MFCD00022870
N-Oxydiethylene-2-benzothiazolesulfenamide
4-(1,3-benzothiazol-2-ylsulfanyl)morpholine
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0897

RTECSNumber : DL5950000

TSCA : Yes

Documents