Learn More
2-(Morpholinosulfonyl)benzylamine hydrochloride, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 918812-18-7 |
---|---|
Molecular Formula | C11H17ClN2O3S |
Molecular Weight (g/mol) | 292.778 |
MDL Number | MFCD09284586 |
InChI Key | RMMDNVBLYNZQFP-UHFFFAOYSA-N |
Synonym | 2-morpholinosulfonyl benzylamine hydrochloride, 1-2-morpholin-4-ylsulfonyl phenyl methanamine hydrochloride, 2-morpholine-4-sulfonyl phenyl methanamine hydrochloride, 1-2-morpholine-4-sulfonyl phenyl methanamine hydrochloride, 2-morpholinosulfonyl phenyl methanamine hydrochloride, 2-morpholinosulfonyl phenyl methanamine hydrochloride salt, 1-2-morpholine-4-sulfonyl phenyl methanamine-hydrogen chloride 1/1 |
PubChem CID | 16320333 |
IUPAC Name | (2-morpholin-4-ylsulfonylphenyl)methanamine;hydrochloride |
SMILES | C1COCCN1S(=O)(=O)C2=CC=CC=C2CN.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC69663CB
|
Thermo Scientific Chemicals
CC69663CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC69663DA
|
Thermo Scientific Chemicals
CC69663DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
918812-18-7 | |
292.778 | |
RMMDNVBLYNZQFP-UHFFFAOYSA-N | |
16320333 | |
C1COCCN1S(=O)(=O)C2=CC=CC=C2CN.Cl |
C11H17ClN2O3S | |
MFCD09284586 | |
2-morpholinosulfonyl benzylamine hydrochloride, 1-2-morpholin-4-ylsulfonyl phenyl methanamine hydrochloride, 2-morpholine-4-sulfonyl phenyl methanamine hydrochloride, 1-2-morpholine-4-sulfonyl phenyl methanamine hydrochloride, 2-morpholinosulfonyl phenyl methanamine hydrochloride, 2-morpholinosulfonyl phenyl methanamine hydrochloride salt, 1-2-morpholine-4-sulfonyl phenyl methanamine-hydrogen chloride 1/1 | |
(2-morpholin-4-ylsulfonylphenyl)methanamine;hydrochloride |
Specifications
918812-18-7 | |
100.0 | |
Amber Glass Bottle | |
MFCD09284586 | |
2-morpholinosulfonyl benzylamine hydrochloride, 1-2-morpholin-4-ylsulfonyl phenyl methanamine hydrochloride, 2-morpholine-4-sulfonyl phenyl methanamine hydrochloride, 1-2-morpholine-4-sulfonyl phenyl methanamine hydrochloride, 2-morpholinosulfonyl phenyl methanamine hydrochloride, 2-morpholinosulfonyl phenyl methanamine hydrochloride salt, 1-2-morpholine-4-sulfonyl phenyl methanamine-hydrogen chloride 1/1 | |
C1COCCN1S(=O)(=O)C2=CC=CC=C2CN.Cl | |
292.778 | |
292.79 | |
2-(Morpholinosulfonyl)benzylamine hydrochloride |
96.5 | |
97% | |
C11H17ClN2O3S | |
250mg | |
RMMDNVBLYNZQFP-UHFFFAOYSA-N | |
(2-morpholin-4-ylsulfonylphenyl)methanamine;hydrochloride | |
16320333 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning