missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Morpholinoethyl Methacrylate (stabilized with MEHQ), TCI America™

$84.03 - $249.03

Chemical Identifiers

CAS 2997-88-8
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD00080577
InChI Key MNZNJOQNLFEAKG-UHFFFAOYSA-N
Synonym Methacrylic Acid 2-Morpholinoethyl Ester
PubChem CID 76343
IUPAC Name 2-morpholin-4-ylethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCN1CCOCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M29011G View Documents TCI America
M29011G
1g Each for $84.03
 
M29015G View Documents TCI America
M29015G
5g Each for $249.03
 
Specifications

Chemical Identifiers

2997-88-8
199.25
MNZNJOQNLFEAKG-UHFFFAOYSA-N
76343
CC(=C)C(=O)OCCN1CCOCC1
C10H17NO3
MFCD00080577
Methacrylic Acid 2-Morpholinoethyl Ester
2-morpholin-4-ylethyl 2-methylprop-2-enoate

Specifications

81°C
1g
C10H17NO3
Methacrylic Acid 2-Morpholinoethyl Ester
CC(=C)C(=O)OCCN1CCOCC1
199.25
199.25
≥98.0% (GC)
Red
2997-88-8
MFCD00080577
MNZNJOQNLFEAKG-UHFFFAOYSA-N
2-morpholin-4-ylethyl 2-methylprop-2-enoate
76343
Clear Liquid at 20°C
2-Morpholinoethyl Methacrylate (stabilized with MEHQ)
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0866

TSCA : No