missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Morpholinoethanesulfonic Acid, TCI America™

$35.14 - $171.11

Chemical Identifiers

CAS 4432-31-9
Molecular Formula C6H13NO4S
Molecular Weight (g/mol) 195.233
MDL Number MFCD00006181
InChI Key SXGZJKUKBWWHRA-UHFFFAOYSA-N
Synonym 2-morpholinoethanesulfonic acid, 4-morpholineethanesulfonic acid, 2-n-morpholino ethanesulfonic acid, mes, 4-morpholinethanesulfonic acid, 2-4-morpholinyl ethanesulfonic acid, morpholineethanesulfonic acid, mes buffering agent, 2-morpholinoethanesulphonic acid, morpholino-n-ethylsulfonate
PubChem CID 78165
ChEBI CHEBI:39005
IUPAC Name 2-morpholin-4-ylethanesulfonic acid
SMILES C1COCCN1CCS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M060625G View Documents TCI America
M060625G
25g Each for $35.14
 
M0606250G View Documents TCI America
M0606250G
250g Each for $171.11
 
Specifications

Chemical Identifiers

4432-31-9
195.233
SXGZJKUKBWWHRA-UHFFFAOYSA-N
78165
2-morpholin-4-ylethanesulfonic acid
C6H13NO4S
MFCD00006181
2-morpholinoethanesulfonic acid, 4-morpholineethanesulfonic acid, 2-n-morpholino ethanesulfonic acid, mes, 4-morpholinethanesulfonic acid, 2-4-morpholinyl ethanesulfonic acid, morpholineethanesulfonic acid, mes buffering agent, 2-morpholinoethanesulphonic acid, morpholino-n-ethylsulfonate
CHEBI:39005
C1COCCN1CCS(=O)(=O)O

Specifications

White
25g
C6H13NO4S
2-morpholinoethanesulfonic acid, 4-morpholineethanesulfonic acid, 2-n-morpholino ethanesulfonic acid, mes, 4-morpholinethanesulfonic acid, 2-4-morpholinyl ethanesulfonic acid, morpholineethanesulfonic acid, mes buffering agent, 2-morpholinoethanesulphonic acid, morpholino-n-ethylsulfonate
C1COCCN1CCS(=O)(=O)O
195.233
CHEBI:39005
Crystal-Powder at 20°C
2-Morpholinoethanesulfonic Acid
316°C
4432-31-9
MFCD00006181
SXGZJKUKBWWHRA-UHFFFAOYSA-N
2-morpholin-4-ylethanesulfonic acid
78165
195.23
≥99.0% (T)
Safety and Handling

Safety and Handling

RTECSNumber : QE3479500

TSCA : Yes