missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Morpholinobenzylamine, 97%, Thermo Scientific™

Manufacturer:  Fisher Scientific Company CC24313DA

 View more versions of this product

Catalog No. CC24313DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Morpholinobenzylamine
96.5
95%
MFCD03086183
1-2-morpholin-4-yl phenyl methanamine, 2-4-morpholinyl phenyl methanamine, 2-morpholin-4-yl benzylamine, 2-morpholin-4-yl phenyl methanamine, 2-morpholin-4-yl-benzylamine, 2-morpholin-4-ylbenzylamine, 2-morpholin-4-ylphenyl methylamine, 2-morpholino benzylamine, 2-morpholinobenzylamine, 2-morpholinophenyl methanamine
RNYURNUANACIKS-UHFFFAOYSA-N
(2-morpholin-4-ylphenyl)methanamine
2776563
95%
204078-48-8
100.0
C11H16N2O
Amber glass bottle
1g
NCC1=CC=CC=C1N1CCOCC1
192.26
192.26

Chemical Identifiers

204078-48-8
192.26
RNYURNUANACIKS-UHFFFAOYSA-N
2776563
NCC1=CC=CC=C1N1CCOCC1
C11H16N2O
MFCD03086183
1-2-morpholin-4-yl phenyl methanamine, 2-4-morpholinyl phenyl methanamine, 2-morpholin-4-yl benzylamine, 2-morpholin-4-yl phenyl methanamine, 2-morpholin-4-yl-benzylamine, 2-morpholin-4-ylbenzylamine, 2-morpholin-4-ylphenyl methylamine, 2-morpholino benzylamine, 2-morpholinobenzylamine, 2-morpholinophenyl methanamine
(2-morpholin-4-ylphenyl)methanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents
Product Certifications
Promotions

Promotions