Learn More
2-Morpholino-3-pyridinamine, 97%, Maybridge™
Manufacturer: Fisher Scientific Company CC24614DA
Specifications
2-Morpholino-3-pyridinamine | |
1g | |
96.5 | |
97% | |
MFCD03001243 | |
DHYQHRDXKCKJAN-UHFFFAOYSA-N | |
2-morpholin-4-ylpyridin-3-amine | |
2776573 | |
97% |
Amber Glass Bottle | |
51627-47-5 | |
100.0 | |
C9H13N3O | |
2-morpholin-4-yl pyridin-3-amine, 2-morpholin-4-yl-3-pyridylamine, 2-morpholin-4-yl-pyridin-3-ylamine, 2-morpholino-3-pyridinamine, 2-morpholinopyridin-3-amine, 3-amino-2-morpholinopyridine, 3-pyridinamine,2-4-morpholinyl | |
C1COCCN1C2=C(C=CC=N2)N | |
179.223 | |
179.22 |
Chemical Identifiers
51627-47-5 | |
179.223 | |
DHYQHRDXKCKJAN-UHFFFAOYSA-N | |
2776573 | |
C1COCCN1C2=C(C=CC=N2)N |
C9H13N3O | |
MFCD03001243 | |
2-morpholin-4-yl pyridin-3-amine, 2-morpholin-4-yl-3-pyridylamine, 2-morpholin-4-yl-pyridin-3-ylamine, 2-morpholino-3-pyridinamine, 2-morpholinopyridin-3-amine, 3-amino-2-morpholinopyridine, 3-pyridinamine,2-4-morpholinyl | |
2-morpholin-4-ylpyridin-3-amine |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning