Please login to your online account to display your discounted pricing

2-Morpholino-1,3-thiazole-5-carboxylic acid, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC62401DA

 View more versions of this product

Catalog No. CC62401DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 214.24
Quantity 1g
Assay 97%
Chemical Name or Material 2-morpholino-1, 3-thiazole-5-carboxylic acid
Assay Percent Range 97%
Molecular Formula C8H10N2O3S
CAS 332345-29-6
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material 2-morpholino-1, 3-thiazole-5-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H10N2O3S
Formula Weight 214.24
Accessories

GHS Signal Word: Warning