Please login to your online account to display your discounted pricing

2-Morpholino-1,3-thiazole-5-carboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-morpholino-1, 3-thiazole-5-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H10N2O3S
Formula Weight 214.24
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC62401CB

 
fisher scientific company
CC62401CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC62401DA

 
fisher scientific company
CC62401DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC62401DE

 
fisher scientific company
CC62401DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 214.24
Quantity 250mg
Assay 97%
Chemical Name or Material 2-morpholino-1, 3-thiazole-5-carboxylic acid
Assay Percent Range 97%
Molecular Formula C8H10N2O3S
CAS 332345-29-6
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Warning
Safety and Handling

GHS Signal Word: Warning