Please login to your online account to display your discounted pricing

2-Morpholino-1,3-thiazole-4-carboxylic acid, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC62801CB

 View more versions of this product

Catalog No. CC62801CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 214.24
Quantity 250mg
Assay 97%
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H10N2O3S
CAS 906353-04-6
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Morpholino-1, 3-thiazole-4-carboxylic acid
MDL Number MFCD09702410
Packaging Amber Glass Bottle
CAS Min % 96.5
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Morpholino-1, 3-thiazole-4-carboxylic acid
CAS 906353-04-6
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H10N2O3S
Formula Weight 214.24
MDL Number MFCD09702410
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.