Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylvaleryl chloride, 98%

Chemical Identifiers

Chemical Name or Material 2-Methylvaleryl chloride
CAS 5238-27-7
Molecular Formula C6H11ClO
Formula Weight 134.61
MDL Number MFCD00013656
UN Number UN2920
Beilstein 1743993
Synonym 2-Methylpentanoyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0272506

  SDS
alfa aesar™
L0272506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0272514

  SDS
alfa aesar™
L0272514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 134.61
Chemical Name or Material 2-Methylvaleryl chloride
Molecular Formula C6H11ClO
MDL Number MFCD00013656
CAS 5238-27-7
Assay 98%
UN Number UN2920
Synonym 2-Methylpentanoyl chloride
Health Hazard 1 Danger
Boiling Point 140° to 144°C
Flash Point 37°C (98°F)
Odor Acrid
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P308+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
Beilstein 1743993
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P308+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.