Learn More
2-Methylthiophene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 127851000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Methylthiophene | |
97.5 | |
-63°C | |
1.0200g/mL | |
7°C | |
98% | |
C5H6S | |
MFCD00005451 | |
17, 37 | |
thiophene, 2-methyl, 2-methyl thiophene, thiophene, methyl, alpha-methylthiophene, ccris 2936, 2-methyl-thiophene, unii-7115jap77a, 2-methylthiacyclopentadiene, a-thiotolene, pubchem5204 | |
XQQBUAPQHNYYRS-UHFFFAOYSA-N | |
2-methylthiophene | |
11126 | |
98% |
554-14-3 | |
100.0 | |
Colorless to Yellow | |
113°C | |
Authentic | |
Glass bottle | |
1.5190 to 1.521 | |
100 g | |
1.02 | |
Solubility in water: immiscible | |
CC1=CC=CS1 | |
98.16 | |
98.16 | |
Liquid |
Chemical Identifiers
554-14-3 | |
98.16 | |
XQQBUAPQHNYYRS-UHFFFAOYSA-N | |
11126 | |
CC1=CC=CS1 |
C5H6S | |
MFCD00005451 | |
thiophene, 2-methyl, 2-methyl thiophene, thiophene, methyl, alpha-methylthiophene, ccris 2936, 2-methyl-thiophene, unii-7115jap77a, 2-methylthiacyclopentadiene, a-thiotolene, pubchem5204 | |
2-methylthiophene |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 209-063-
RUO – Research Use Only