missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methylthio)thiazole, 98%

$261.68 - $768.09

Chemical Identifiers

CAS 5053-24-7
Molecular Formula C4H5NS2
Formula Weight 131.21
MDL Number MFCD00858981
Synonym 2-(Methylmercapto)thiazole
Chemical Name or Material 2-(Methylthio)thiazole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2513714 SDS Alfa Aesar™
B2513714
25g Each for $261.68
AAB2513722 SDS Alfa Aesar™
B2513722
100g Each for $768.09
AAB2513706 SDS Alfa Aesar™
B2513706
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

5053-24-7
131.21
2-(Methylmercapto)thiazole
C4H5NS2
MFCD00858981
2-(Methylthio)thiazole

Specifications

90°C (195°F)
205° to 207°C
5053-24-7
131.21
2-(Methylmercapto)thiazole
98%
Characteristic
25g
C4H5NS2
MFCD00858981
2-(Methylthio)thiazole

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning