Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methylthio)pyridine-3-boronic acid, 95%

$323.24 - $735.77

Chemical Identifiers

Chemical Name or Material 2-(Methylthio)pyridine-3-boronic acid
CAS 1072944-21-8
Molecular Formula C6H8BNO2S
Formula Weight 169.01
MDL Number MFCD09037488
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2716203

 
alfa aesar™
H2716203
1g Each for $735.77

AAH27162MD

 
alfa aesar™
H27162MD
250mg Each for $323.24
Description & Specifications

Specifications

Formula Weight 169.01
Chemical Name or Material 2-(Methylthio)pyridine-3-boronic acid
Assay 95%
Molecular Formula C6H8BNO2S
MDL Number MFCD09037488
CAS 1072944-21-8
Health Hazard 1 Warning
Melting Point ≈100°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.