missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methylthio)phenyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ L1142106

 View more versions of this product

Catalog No. AAL1142106



Specifications

Specifications

2-(Methylthio)phenyl isothiocyanate
149°C to 150°C (10mmHg)
51333-75-6
MFCD00041061
3243502
SIODOISHDGRELW-UHFFFAOYSA-N
1-isothiocyanato-2-methylsulfanylbenzene
142840
97%
>110°C (230°F)
5g
C8H7NS2
UN2922
2-methylthio phenyl isothiocyanate, 1-isothiocyanato-2-methylsulfanyl benzene, acmc-20aomj, 2-methylthiobenzenisothiocyanate, 2-methylthio-phenyl-isothiocyanate, 1-isothiocyanato-2-methylthio benzene, 2-methylthio phenylisothiocyanate, 1-isothiocyanato-2-methylsulfanyl-benzene, benzene,1-isothiocyanato-2-methylthio, 2-methylthio phenyl isothiocyanate
CSC1=CC=CC=C1N=C=S
181.271
181.28

Chemical Identifiers

51333-75-6
181.271
SIODOISHDGRELW-UHFFFAOYSA-N
142840
CSC1=CC=CC=C1N=C=S
C8H7NS2
MFCD00041061
2-methylthio phenyl isothiocyanate, 1-isothiocyanato-2-methylsulfanyl benzene, acmc-20aomj, 2-methylthiobenzenisothiocyanate, 2-methylthio-phenyl-isothiocyanate, 1-isothiocyanato-2-methylthio benzene, 2-methylthio phenylisothiocyanate, 1-isothiocyanato-2-methylsulfanyl-benzene, benzene,1-isothiocyanato-2-methylthio, 2-methylthio phenyl isothiocyanate
1-isothiocyanato-2-methylsulfanylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Documents