missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methylthio)oxazolo[5,4-c]pyridine

$77.42 - $269.38

Chemical Identifiers

CAS 169205-96-3
Molecular Formula C7H6N2OS
Formula Weight 166.2
MDL Number MFCD08447111
Chemical Name or Material 2-(Methylthio)oxazolo[5,4-c]pyridine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5101703 SDS Alfa Aesar™
H5101703
1g Each for $269.38
AAH51017MD SDS Alfa Aesar™
H51017MD
250mg Each for $77.42
Specifications Description & Specifications

Chemical Identifiers

169205-96-3
166.2
2-(Methylthio)oxazolo[5,4-c]pyridine
C7H6N2OS
MFCD08447111

Specifications

169205-96-3
166.2
79° to 81°C
2-(Methylthio)oxazolo[5,4-c]pyridine
C7H6N2OS
MFCD08447111
1g

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning