Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methylthio)nicotinoyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 2-(Methylthio)nicotinoyl chloride
CAS 97936-43-1
Molecular Formula C7H6ClNOS
Formula Weight 187.65
MDL Number MFCD00051678
UN Number UN3261
Beilstein 5506864
Synonym 2-(Methylmercapto)nicotinoyl chloride; 2-(Methylmercapto)pyridine-3-carbonyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1791606

 
alfa aesar™
A1791606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1791603

 
alfa aesar™
A1791603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1791614

 
alfa aesar™
A1791614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 187.65
Chemical Name or Material 2-(Methylthio)nicotinoyl chloride
Assay 98%
Molecular Formula C7H6ClNOS
MDL Number MFCD00051678
CAS 97936-43-1
UN Number UN3261
Beilstein 5506864
Synonym 2-(Methylmercapto)nicotinoyl chloride; 2-(Methylmercapto)pyridine-3-carbonyl chloride
Health Hazard 1 Danger
Melting Point 92° to 96°C
Quantity 5g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.