missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Methylthio)ethanol, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC210101000

 View more versions of this product

Catalog No. AC210101000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-(Methylthio)ethanol
70°C
Glass bottle
1.06
Colorless
5271-38-5
100.0
C3H8OS
MFCD00002908
2-hydroxyethyl methyl sulfide
WBBPRCNXBQTYLF-UHFFFAOYSA-N
2-methylsulfanylethanol
78925
92.15
≥98.5%
1.0600g/mL
Authentic
1.4934 to 1.4954
169°C to 171°C
100mL
98.5
99%
CH3SCH2CH2OH
01, II, 524
Solubility in water: insoluble
CSCCO
92.15
CHEBI:63861
Liquid

Chemical Identifiers

5271-38-5
92.15
WBBPRCNXBQTYLF-UHFFFAOYSA-N
78925
2-methylsulfanylethanol
C3H8OS
MFCD00002908
2-hydroxyethyl methyl sulfide
CHEBI:63861
CSCCO
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents