missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Methylthio)benzothiazole 98.0+%, TCI America™
Supplier: TCI America M043925G
Specifications
2-(Methylthio)benzothiazole | |
46°C | |
177°C | |
MFCD00005784 | |
2-methylthio benzothiazole, 2-methylmercaptobenzothiazole, 2-methylthio benzo d thiazole, 2-methylthiobenzothiazole, 2-methylmercapto benzothiazole, benzothiazole, 2-methylthio, mtbt, 2-methylthio-1,3-benzothiazole, 2-methylsulfanyl-1,3-benzothiazole, 2-methyl mercaptobenzothiazole | |
CSC1=NC2=CC=CC=C2S1 | |
181.271 | |
CHEBI:1217 | |
≥98.0% (GC) |
615-22-5 | |
White-Yellow | |
C8H7NS2 | |
25 g | |
UTBVIMLZIRIFFR-UHFFFAOYSA-N | |
2-methylsulfanyl-1,3-benzothiazole | |
11989 | |
181.27 | |
Crystalline Powder |
Chemical Identifiers
615-22-5 | |
181.271 | |
UTBVIMLZIRIFFR-UHFFFAOYSA-N | |
11989 | |
2-methylsulfanyl-1,3-benzothiazole |
C8H7NS2 | |
MFCD00005784 | |
2-methylthio benzothiazole, 2-methylmercaptobenzothiazole, 2-methylthio benzo d thiazole, 2-methylthiobenzothiazole, 2-methylmercapto benzothiazole, benzothiazole, 2-methylthio, mtbt, 2-methylthio-1,3-benzothiazole, 2-methylsulfanyl-1,3-benzothiazole, 2-methyl mercaptobenzothiazole | |
CHEBI:1217 | |
CSC1=NC2=CC=CC=C2S1 |
Safety and Handling
EINECSNumber : (5)-0244
TSCA : Yes