missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Methylthio)benzonitrile, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0687003

 View more versions of this product

Catalog No. AAL0687003



Description

Description

2-(Methylthio)benzonitrile is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)benzonitrile is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2-(Methylthio)benzonitrile
33°C to 38°C
C8H7NS
1 g
2207135
Insoluble in water.
CSC1=CC=CC=C1C#N
149.211
149.22
6609-54-7
116°C to 117°C (5 mmHg)
MFCD00015557
UN3439
2-methylthio benzonitrile, 2-methylsulfanyl benzonitrile, 2-cyanophenyl methyl sulfide, 2-methylmercapto benzonitrile, methylthio benzonitrile, 2-methylthiobenzenecarbonitrile, benzonitrile,2-methylthio, 2-methylsulfanyl benzenecarbonitrile
PXZSANDJGNKIIA-UHFFFAOYSA-N
2-methylsulfanylbenzonitrile
138781
98%

Chemical Identifiers

6609-54-7
149.211
PXZSANDJGNKIIA-UHFFFAOYSA-N
138781
CSC1=CC=CC=C1C#N
C8H7NS
MFCD00015557
2-methylthio benzonitrile, 2-methylsulfanyl benzonitrile, 2-cyanophenyl methyl sulfide, 2-methylmercapto benzonitrile, methylthio benzonitrile, 2-methylthiobenzenecarbonitrile, benzonitrile,2-methylthio, 2-methylsulfanyl benzenecarbonitrile
2-methylsulfanylbenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only