Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methylthio)benzimidazole, 98+%

$36.01 - $577.09

Chemical Identifiers

Chemical Name or Material 2- (Methylthio)benzimidazole
CAS 7152-24-1
Molecular Formula C8H8N2S
Formula Weight 164.23
MDL Number MFCD00005594
Beilstein 122360
Synonym 2-(Methylmercapto)benzimidazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1068609

  SDS
alfa aesar™
A1068609
10g Each for $36.01

AAA1068618

  SDS
alfa aesar™
A1068618
50g Each for $138.62

AAA1068630

  SDS
alfa aesar™
A1068630
250g Each for $577.09
Description & Specifications

Specifications

Formula Weight 164.23
Chemical Name or Material 2- (Methylthio)benzimidazole
TSCA TSCA
Assay ≥98%
Molecular Formula C8H8N2S
MDL Number MFCD00005594
CAS 7152-24-1
Beilstein 122360
Synonym 2-(Methylmercapto)benzimidazole
Melting Point 203° to 205°C
Quantity 10g
RTECS Number DD9480000