missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methylthio)benzeneboronic acid, 98+%

$36.09 - $91.76

Chemical Identifiers

CAS 168618-42-6
Molecular Formula C7H9BO2S
Molecular Weight (g/mol) 168.017
MDL Number MFCD01318165
InChI Key QXBWTYBCNFKURT-UHFFFAOYSA-N
Synonym 2-methylthio phenylboronic acid, 2-methylthiophenylboronic acid, 2-methylthio phenyl boronic acid, 2-methylthio benzeneboronic acid, 2-thioanisoleboronic acid, 2-methylsulfanylphenyl boronic acid, 2-methylsulfanyl phenylboronic acid, 2-methylsulfanyl phenyl boranediol, 2-methylsulfanyl phenyl boronic acid, o-methylthio phenylboronic acid
PubChem CID 2773543
IUPAC Name (2-methylsulfanylphenyl)boronic acid
SMILES B(C1=CC=CC=C1SC)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1745606 View Documents Alfa Aesar™
L1745606
5g Each for $91.76
 
AAL1745603 View Documents Alfa Aesar™
L1745603
1g Each for $36.09
 
Specifications

Chemical Identifiers

168618-42-6
168.017
QXBWTYBCNFKURT-UHFFFAOYSA-N
2773543
B(C1=CC=CC=C1SC)(O)O
C7H9BO2S
MFCD01318165
2-methylthio phenylboronic acid, 2-methylthiophenylboronic acid, 2-methylthio phenyl boronic acid, 2-methylthio benzeneboronic acid, 2-thioanisoleboronic acid, 2-methylsulfanylphenyl boronic acid, 2-methylsulfanyl phenylboronic acid, 2-methylsulfanyl phenyl boranediol, 2-methylsulfanyl phenyl boronic acid, o-methylthio phenylboronic acid
(2-methylsulfanylphenyl)boronic acid

Specifications

~160°C
168618-42-6
MFCD01318165
2-methylthio phenylboronic acid, 2-methylthiophenylboronic acid, 2-methylthio phenyl boronic acid, 2-methylthio benzeneboronic acid, 2-thioanisoleboronic acid, 2-methylsulfanylphenyl boronic acid, 2-methylsulfanyl phenylboronic acid, 2-methylsulfanyl phenyl boranediol, 2-methylsulfanyl phenyl boronic acid, o-methylthio phenylboronic acid
B(C1=CC=CC=C1SC)(O)O
168.017
168.02
2-(Methylthio)benzeneboronic acid
5g
C7H9BO2S
8309529
QXBWTYBCNFKURT-UHFFFAOYSA-N
(2-methylsulfanylphenyl)boronic acid
2773543
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning