Learn More
2-(Methylthio)aniline, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 127110250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-(Methylthio)aniline | |
96.0 | |
1.1110g/mL | |
>110°C | |
96% min. (GC) | |
C7H9NS | |
CH3SC6H4NH2 | |
25g | |
1.111 | |
WBRPQQSADOCKCH-UHFFFAOYSA-N | |
2-methylsulfanylaniline | |
76337 | |
97% |
2987-53-3 | |
100.0 | |
234.0°C | |
Authentic | |
Glass bottle | |
1.6230 to 1.6250 (20°C, 589nm) | |
MFCD00007708 | |
13, 399 | |
2-methylthio aniline, 2-aminothioanisole, 2-methylsulfanyl aniline, 2-methylmercaptoaniline, 2-methylmercapto aniline, o-thioanisidine, o-aminothioanisole, 2-amino thioanisole, benzenamine, 2-methylthio, o-methylthio aniline | |
CSC1=CC=CC=C1N | |
139.22 | |
139.22 |
Chemical Identifiers
2987-53-3 | |
139.22 | |
WBRPQQSADOCKCH-UHFFFAOYSA-N | |
76337 | |
CSC1=CC=CC=C1N |
C7H9NS | |
MFCD00007708 | |
2-methylthio aniline, 2-aminothioanisole, 2-methylsulfanyl aniline, 2-methylmercaptoaniline, 2-methylmercapto aniline, o-thioanisidine, o-aminothioanisole, 2-amino thioanisole, benzenamine, 2-methylthio, o-methylthio aniline | |
2-methylsulfanylaniline |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
GHS Signal Word: Danger
EINECSNumber : 221-055-9
RUO â Research Use Only