missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Methylthio)-6-(trifluoromethyl)pyrimidin-4(1H)-one, TCI America™

$64.17 - $189.45

Chemical Identifiers

CAS 16097-62-4
Molecular Formula C6H5F3N2OS
Molecular Weight (g/mol) 210.174
MDL Number MFCD00153191
InChI Key BRLPDRZKOMZVOY-UHFFFAOYSA-N
Synonym 2-methylthio-6-trifluoromethyl pyrimidin-4-ol, 4-hydroxy-2-methylthio-6-trifluoromethyl pyrimidine, 2-methylsulfanyl-6-trifluoromethyl pyrimidin-4-ol, 2-methylsulfanyl-6-trifluoromethyl-4-pyrimidinol, methylsulfanyltrifluoromethylpyrimidinol, 2-methylsulfanyl-6-trifluoromethyl-1h-pyrimidin-4-one, 4-hydroxy-2-methylthio-6-trifluoromethyl-pyrimidine, pubchem16996, 2-methylthio-4-trifluoromethyl-6-pyrimidinol, 2-methylthio-6-trifluoromethylpyrimidin-4-ol
PubChem CID 248062
IUPAC Name 2-methylsulfanyl-6-(trifluoromethyl)-1H-pyrimidin-4-one
SMILES CSC1=NC(=O)C=C(N1)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M28515G View Documents TCI America
M28515G
5g Each for $189.45
 
M28511G View Documents TCI America
M28511G
1g Each for $64.17
 
Specifications

Chemical Identifiers

16097-62-4
210.174
BRLPDRZKOMZVOY-UHFFFAOYSA-N
248062
CSC1=NC(=O)C=C(N1)C(F)(F)F
C6H5F3N2OS
MFCD00153191
2-methylthio-6-trifluoromethyl pyrimidin-4-ol, 4-hydroxy-2-methylthio-6-trifluoromethyl pyrimidine, 2-methylsulfanyl-6-trifluoromethyl pyrimidin-4-ol, 2-methylsulfanyl-6-trifluoromethyl-4-pyrimidinol, methylsulfanyltrifluoromethylpyrimidinol, 2-methylsulfanyl-6-trifluoromethyl-1h-pyrimidin-4-one, 4-hydroxy-2-methylthio-6-trifluoromethyl-pyrimidine, pubchem16996, 2-methylthio-4-trifluoromethyl-6-pyrimidinol, 2-methylthio-6-trifluoromethylpyrimidin-4-ol
2-methylsulfanyl-6-(trifluoromethyl)-1H-pyrimidin-4-one

Specifications

White
5g
C6H5F3N2OS
2-methylthio-6-trifluoromethyl pyrimidin-4-ol, 4-hydroxy-2-methylthio-6-trifluoromethyl pyrimidine, 2-methylsulfanyl-6-trifluoromethyl pyrimidin-4-ol, 2-methylsulfanyl-6-trifluoromethyl-4-pyrimidinol, methylsulfanyltrifluoromethylpyrimidinol, 2-methylsulfanyl-6-trifluoromethyl-1h-pyrimidin-4-one, 4-hydroxy-2-methylthio-6-trifluoromethyl-pyrimidine, pubchem16996, 2-methylthio-4-trifluoromethyl-6-pyrimidinol, 2-methylthio-6-trifluoromethylpyrimidin-4-ol
CSC1=NC(=O)C=C(N1)C(F)(F)F
210.174
210.17
≥98.0% (GC)
180°C
16097-62-4
MFCD00153191
BRLPDRZKOMZVOY-UHFFFAOYSA-N
2-methylsulfanyl-6-(trifluoromethyl)-1H-pyrimidin-4-one
248062
Crystal-Powder at 20°C
2-(Methylthio)-6-(trifluoromethyl)pyrimidin-4(1H)-one
Safety and Handling

Safety and Handling

TSCA : No