Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylthio-5-(trifluoromethyl)aniline, 97%

Chemical Identifiers

Chemical Name or Material 2-Methylthio-5-(trifluoromethyl)aniline
CAS 207974-07-0
Molecular Formula C8H8F3NS
Formula Weight 207.22
MDL Number MFCD00052715
UN Number UN2810
Beilstein 9122593
Synonym 3-Amino-4-(methylthio)benzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1145203

  SDS
alfa aesar™
L1145203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 207.22
Molecular Formula C8H8F3NS
MDL Number MFCD00052715
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Methylthio-5-(trifluoromethyl)aniline
Assay 97%
CAS 207974-07-0
UN Number UN2810
Synonym 3-Amino-4-(methylthio)benzotrifluoride
Health Hazard 1 Danger
Boiling Point 111° to 112°C (12mmHg)
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Beilstein 9122593
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.