Please login to your online account to display your discounted pricing

2-(Methylthio)-4,5-dihydro-1H-imidazole hydrobromide, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(Methylthio)-4, 5-dihydro-1H-imidazole hydrobromide
CAS 40241-78-9
CAS Min % 80.0
CAS Max % 100.0
Assay Tech.
Assay Percent Range 95%
Molecular Formula C4H8N2S·BrH
Formula Weight 197.1
MDL Number MFCD01570110
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DP01481DA

 
fisher scientific company
DP01481DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Technical Grade
Formula Weight 197.1
Quantity 1g
Assay Tech.
CAS Max % 100.0
CAS Min % 80.0
Chemical Name or Material 2-(Methylthio)-4, 5-dihydro-1H-imidazole hydrobromide
Molecular Formula C4H8N2S·BrH
CAS 40241-78-9
Packaging Amber Glass Bottle
Assay Percent Range 95%
MDL Number MFCD01570110
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.