Please login to your online account to display your discounted pricing

2-(Methylthio)-4,5-dihydro-1H-imidazole hydrobromide, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  DP01481DA

 View more versions of this product

Catalog No. DP01481DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 197.1
Purity Grade Technical Grade
Quantity 1g
Assay Tech.
CAS Max % 100.0
CAS Min % 80.0
Chemical Name or Material 2-(Methylthio)-4, 5-dihydro-1H-imidazole hydrobromide
Molecular Formula C4H8N2S·BrH
CAS 40241-78-9
MDL Number MFCD01570110
Packaging Amber Glass Bottle
Assay Percent Range 95%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material 2-(Methylthio)-4, 5-dihydro-1H-imidazole hydrobromide
CAS 40241-78-9
CAS Min % 80.0
CAS Max % 100.0
Assay Tech.
Assay Percent Range 95%
Molecular Formula C4H8N2S·BrH
Formula Weight 197.1
MDL Number MFCD01570110
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents