missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[2-[(Methylthio)(2H-pyrrol-2-ylidene)methyl]-1H-pyrrole](difluoroborane), TCI America™

$245.98 - $737.94

Chemical Identifiers

CAS 892505-41-8
Molecular Formula C10H9BF2N2S
Molecular Weight (g/mol) 238.063
InChI Key OSUYHKLOJHHXAI-UHFFFAOYSA-N
PubChem CID 23631051
SMILES [B-]1(N2C=CC=C2C(=C3[N+]1=CC=C3)SC)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M2609200MG View Documents TCI America
M2609200MG
200mg Each for $245.98
 
M26091G View Documents TCI America
M26091G
1g Each for $737.94
 
Specifications

Chemical Identifiers

892505-41-8
238.063
23631051
C10H9BF2N2S
OSUYHKLOJHHXAI-UHFFFAOYSA-N
[B-]1(N2C=CC=C2C(=C3[N+]1=CC=C3)SC)(F)F

Specifications

Red
200mg
C10H9BF2N2S
[B-]1(N2C=CC=C2C(=C3[N+]1=CC=C3)SC)(F)F
23631051
Crystal-Powder at 20°C
[2-[(Methylthio)(2H-pyrrol-2-ylidene)methyl]-1H-pyrrole](difluoroborane)
110°C
892505-41-8
OSUYHKLOJHHXAI-UHFFFAOYSA-N
238.063
238.06
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No