Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyltetrahydrothiophen-3-one, 97%

$212.46 - $212.46

Chemical Identifiers

Chemical Name or Material 2-Methyltetrahydrothiophen-3-one
CAS 13679-85-1
Molecular Formula C5H8OS
Formula Weight 116.18
MDL Number MFCD00078280
Beilstein 106443
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1008306

  SDS
alfa aesar™
L1008306
5g Each for $212.46
Description & Specifications

Specifications

Formula Weight 116.18
Chemical Name or Material 2-Methyltetrahydrothiophen-3-one
Assay 97%
MDL Number MFCD00078280
CAS 13679-85-1
Beilstein 106443
Health Hazard 1 Warning
Quantity 5g
Flash Point 69°C (156°F)
Health Hazard 2 GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P264-P301+P312-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Molecular Formula C5H8OS
Boiling Point 83° to 84°C (18mmHg)
Safety and Handling

Warning

GHS P Statement
P210-P280-P264-P301+P312-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.