missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyltetrahydrothiophen-3-one, 97%

Manufacturer:  Alfa Aesar™ L1008306

 View more versions of this product

Catalog No. AAL1008306



Specifications

Specifications

2-Methyltetrahydrothiophen-3-one
83°C to 84°C (18mmHg)
13679-85-1
MFCD00078280
2-methyltetrahydrothiophen-3-one, dihydro-2-methyl-3 2h-thiophenone, 2-methyldihydrothiophen-3 2h-one, 3 2h-thiophenone, dihydro-2-methyl, 2-methyltetrahydrothiophene-3-one, 2-methyl-3-oxotetrahydrothiophene, 2-methyl-4,5-dihydro-3 2h-thiophenone, 2-methyl-3-tetrahydrothiophenone, 2-methyl tetrahydrothiophen-3-one, xi-dihydro-2-methyl-3 2h-thiophenone
CC1C(=O)CCS1
116.178
CHEBI:87506
97%
69°C (156°F)
5g
C5H8OS
106443
YMZZPMVKABUEBL-UHFFFAOYSA-N
2-methylthiolan-3-one
61664
116.18

Chemical Identifiers

13679-85-1
116.178
YMZZPMVKABUEBL-UHFFFAOYSA-N
61664
2-methylthiolan-3-one
C5H8OS
MFCD00078280
2-methyltetrahydrothiophen-3-one, dihydro-2-methyl-3 2h-thiophenone, 2-methyldihydrothiophen-3 2h-one, 3 2h-thiophenone, dihydro-2-methyl, 2-methyltetrahydrothiophene-3-one, 2-methyl-3-oxotetrahydrothiophene, 2-methyl-4,5-dihydro-3 2h-thiophenone, 2-methyl-3-tetrahydrothiophenone, 2-methyl tetrahydrothiophen-3-one, xi-dihydro-2-methyl-3 2h-thiophenone
CHEBI:87506
CC1C(=O)CCS1
Safety and Handling

Safety and Handling

GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.

GHS P Statement
P210-P280-P264-P301+P312-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents