missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyltetrahydrofuran (stabilized with BHT), TCI America™

Manufacturer:  TCI America M043725ML

 View more versions of this product

Catalog No. M043725ML



Specifications

Specifications

2-Methyltetrahydrofuran (stabilized with BHT)
Colorless
96-47-9
MFCD00005367
2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
CC1CCCO1
86.134
86.13
≥98.0% (GC)
78°C
25mL
C5H10O
2536
JWUJQDFVADABEY-UHFFFAOYSA-N
2-methyloxolane
7301
Clear Liquid at 20°C

Chemical Identifiers

96-47-9
86.134
JWUJQDFVADABEY-UHFFFAOYSA-N
7301
CC1CCCO1
C5H10O
MFCD00005367
2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
2-methyloxolane
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6944

RTECSNumber : LU2800000

TSCA : Yes

Documents