missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilizer free, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC397201000

 View more versions of this product

Catalog No. AC397201000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Methyltetrahydrofuran
100mL
96-47-9
100.0
Authentic
20ppm max. (at lot release)
Extra Dry
≥99.0%
C5H10O
86.13
15, 6198
Clear colorless
Stabilizer free, Extra Dry
99.0
0.8600g/mL
AcroSeal™ Glass Bottle
0.001% max.
Liquid
99+%
0.002% max.
MFCD00005367
2-MeTHF

Chemical Identifiers

96-47-9
100.0
C5H10O
MFCD00005367
2-Methyltetrahydrofuran, Stabilizer free, Extra Dry
Extra Dry
99.0
99+%
86.13
2-MeTHF
≥99.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger