missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyltetrahydrofuran-3-thiol (cis- and trans- mixture) 98.0+%, TCI America™

Manufacturer:  Tci America M18411G

 View more versions of this product

Catalog No. M18411G



Specifications

Specifications

2-Methyltetrahydrofuran-3-thiol (cis- and trans- mixture)
Yellow
57124-87-5
MFCD01632596
2-methyl-3-mercaptotetrahydrofuran, 2-methyl-3-tetrahydrofuran thiol, 2-methyl-3-tetrahydrofuranthiol, 2-methyltetrahydrofuran-3-thiol, 3-furanthiol, tetrahydro-2-methyl, 3-mercapto-2-methyltetrahydrofuran, ksc201q4p, pentitol, 1,4-anhydro-2,5-dideoxy-3-thio, pubchem9972, tetrahydro-2-methyl-3-furanthiol
CC1C(CCO1)S
118.194
118.19
≥98.0% (GC)
168°C
1g
C5H10OS
3336
DBPHPBLAKVZXOY-UHFFFAOYSA-N
2-methyloxolane-3-thiol
62128
Clear Liquid at 20°C

Chemical Identifiers

57124-87-5
118.194
DBPHPBLAKVZXOY-UHFFFAOYSA-N
62128
CC1C(CCO1)S
C5H10OS
MFCD01632596
2-methyl-3-mercaptotetrahydrofuran, 2-methyl-3-tetrahydrofuran thiol, 2-methyl-3-tetrahydrofuranthiol, 2-methyltetrahydrofuran-3-thiol, 3-furanthiol, tetrahydro-2-methyl, 3-mercapto-2-methyltetrahydrofuran, ksc201q4p, pentitol, 1,4-anhydro-2,5-dideoxy-3-thio, pubchem9972, tetrahydro-2-methyl-3-furanthiol
2-methyloxolane-3-thiol
Safety and Handling

Safety and Handling

TSCA : Yes

Documents
Promotions

Promotions