Please login to your online account to display your discounted pricing

2-Methyltetrahydrofuran-3-thiol, 97%, mixed isomers, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  332700010

 View more versions of this product

Catalog No. AC332700010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 118.2
Quantity 1g
Color Clear colorless to pale yellow
Density 1.0400g/mL
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 2-Methyltetrahydrofuran-3-thiol
Name Note Mixture of isomers
Assay Percent Range 97%
Molecular Formula C5H10OS
CAS 57124-87-5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
May cause respiratory irritation.
Infrared Spectrum Authentic
Refractive Index 1.4730 to 1.4910 (20°C, 589nm)
MDL Number MFCD01632596
Health Hazard 3 GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Methyltetrahydrofuran-3-thiol
Name Note Mixture of isomers
CAS 57124-87-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H10OS
Formula Weight 118.2
MDL Number MFCD01632596
Accessories

GHS Signal Word: Warning

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
May cause respiratory irritation.

SDS & Safety Documents