missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyltetrahydrofuran-3-one 98.0+%, TCI America™

$49.38 - $144.84

Chemical Identifiers

CAS 3188-00-9
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00010423
InChI Key FCWYQRVIQDNGBI-UHFFFAOYSA-N
Synonym 2-methyl tetrahydro-3-furanone
PubChem CID 18522
IUPAC Name 2-methyloxolan-3-one
SMILES CC1C(=O)CCO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M10555G View Documents Tci America
M10555G
5g Each for $49.38
 
M105525G View Documents Tci America
M105525G
25g Each for $144.84
 
Specifications

Chemical Identifiers

3188-00-9
100.117
FCWYQRVIQDNGBI-UHFFFAOYSA-N
18522
CC1C(=O)CCO1
C5H8O2
MFCD00010423
2-methyl tetrahydro-3-furanone
2-methyloxolan-3-one

Specifications

139°C
5g
C5H8O2
1224
FCWYQRVIQDNGBI-UHFFFAOYSA-N
2-methyloxolan-3-one
18522
Clear Liquid at 20°C
2-Methyltetrahydrofuran-3-one
Yellow
3188-00-9
MFCD00010423
2-methyl tetrahydro-3-furanone
CC1C(=O)CCO1
100.117
100.12
≥98.0% (GC)
Safety and Handling

Safety and Handling

RTECSNumber : LU3579000

TSCA : Yes