Please login to your online account to display your discounted pricing

2-Methyltetrahydrofuran-3-one, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Methyltetrahydrofuran-3-one
CAS 3188-00-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H8O2
Formula Weight 100.12
MDL Number MFCD00010423
Synonym Dihydro-2-methyl-3(2H)-furanone
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB01113DA

  SDS
fisher scientific company
SB01113DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 100.12
Quantity 1g
Assay 97%
Density 1.045g/mL
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C5H8O2
Synonym Dihydro-2-methyl-3(2H)-furanone
Chemical Name or Material 2-Methyltetrahydrofuran-3-one
Packaging Amber Glass Bottle
MDL Number MFCD00010423
CAS 3188-00-9
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.