missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methylsulfonylamino)benzeneboronic acid, 95%

Manufacturer:  Alfa Aesar™ H5328803

 View more versions of this product

Catalog No. AAH5328803



Specifications

Specifications

2-(Methylsulfonylamino)benzeneboronic acid
1g
C7H10BNO4S
2-methylsulfonamido phenyl boronic acid, 2-methanesulfonylaminophenylboronic acid, 2-methylsulfonylaminophenylboronic acid, 2-methanesulfonamidophenyl boronic acid, 2-methanesulfonylamino phenylboronic acid, 2-methanesulfonamido phenyl boronic acid, 2-methylsulphonylamino benzeneboronic acid, 2-methanesulfonamidophenylboronic acid, 2-methylsulfonylamino benzeneboronic acid, pubchem6370
B(C1=CC=CC=C1NS(=O)(=O)C)(O)O
215.03
215.03
116°C to 120°C
756520-78-2
MFCD02179471
RPRMOEFOTPSWBO-UHFFFAOYSA-N
[2-(methanesulfonamido)phenyl]boronic acid
2773533
95%

Chemical Identifiers

756520-78-2
215.03
RPRMOEFOTPSWBO-UHFFFAOYSA-N
2773533
B(C1=CC=CC=C1NS(=O)(=O)C)(O)O
C7H10BNO4S
MFCD02179471
2-methylsulfonamido phenyl boronic acid, 2-methanesulfonylaminophenylboronic acid, 2-methylsulfonylaminophenylboronic acid, 2-methanesulfonamidophenyl boronic acid, 2-methanesulfonylamino phenylboronic acid, 2-methanesulfonamido phenyl boronic acid, 2-methylsulphonylamino benzeneboronic acid, 2-methanesulfonamidophenylboronic acid, 2-methylsulfonylamino benzeneboronic acid, pubchem6370
[2-(methanesulfonamido)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents