Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methylsulfonyl)thiophene, 97%

$117.38 - $117.38

Chemical Identifiers

Chemical Name or Material 2-(Methylsulfonyl)thiophene
CAS 38695-60-2
Molecular Formula C5H6O2S2
Formula Weight 162.23
MDL Number MFCD00052826
Synonym 2-(Methanesulfonyl)thiophene; Methyl 2-thienyl sulfone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2094003

  SDS
alfa aesar™
B2094003
1g Each for $117.38
Description & Specifications

Specifications

Formula Weight 162.23
Assay 97%
MDL Number MFCD00052826
CAS 38695-60-2
Synonym 2-(Methanesulfonyl)thiophene; Methyl 2-thienyl sulfone
Melting Point 46° to 49°C
Molecular Formula C5H6O2S2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-(Methylsulfonyl)thiophene
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.