Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methylsulfonyl)ethanol, 97%

$63.89 - $63.89

Chemical Identifiers

Chemical Name or Material 2-(Methylsulfonyl)ethanol
CAS 15205-66-0
Molecular Formula C3H8O3S
Formula Weight 124.16
MDL Number MFCD00014747
Beilstein 1746915
Synonym 2-Hydroxyethyl methyl sulfone; 2-(Methanesulfonyl)ethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0315906

  SDS
alfa aesar™
L0315906
5g Each for $63.89
Description & Specifications

Specifications

Formula Weight 124.16
Chemical Name or Material 2-(Methylsulfonyl)ethanol
MDL Number MFCD00014747
CAS 15205-66-0
Assay 97%
Molecular Formula C3H8O3S
Beilstein 1746915
Synonym 2-Hydroxyethyl methyl sulfone; 2-(Methanesulfonyl)ethanol
Melting Point 27° to 30°C
Boiling Point 148° to 149°C (1mmHg)
Quantity 5g
Flash Point >110°C (230°F)

Applications
2-(Methylsulfonyl)ethanol may be used as reagent in the synthesis of phenols from aryl fluorides. It may be used in the preparation of 2-(methylsulfonyl)ethyl N,N-bis(2-chloroethyl)phosphorodiamidate. 2-Hydroxyethyl Methyl Sulfone, is the versatile building block used in the synthesis of more complex sulfur containing compounds. It can be used for the studies of inhibitor and substrate moieties to acetylcholinesterase.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.