Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methylsulfonyl)benzoic acid, 98%

$76.39 - $325.46

Chemical Identifiers

Chemical Name or Material 2-(Methylsulfonyl)benzoic acid
CAS 33963-55-2
Molecular Formula C8H8O4S
Formula Weight 200.21
MDL Number MFCD00216492
Synonym 2-(Methanesulfonyl)benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5101803

  SDS
alfa aesar™
H5101803
1g Each for $76.39

AAH5101806

  SDS
alfa aesar™
H5101806
5g Each for $325.46
Description & Specifications

Specifications

Formula Weight 200.21
Quantity 1g
Molecular Formula C8H8O4S
Assay 98%
Synonym 2-(Methanesulfonyl)benzoic acid
Health Hazard 1 Warning
Chemical Name or Material 2-(Methylsulfonyl)benzoic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 136° to 141°C
MDL Number MFCD00216492
CAS 33963-55-2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.