missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-(Methylsulfonyl)acetanilide, 96%

$28.82 - $335.07

Chemical Identifiers

CAS 20628-27-7
Molecular Formula C9H11NO3S
Molecular Weight (g/mol) 213.251
MDL Number MFCD01464107
InChI Key NGWCPCSZLLJKPD-UHFFFAOYSA-N
Synonym 2'-methylsulfonyl acetanilide, 2'-methylsulfonylacetanilide, acmc-1ceiy, n-2-methanesulfonyl phenyl acetamide, n-2-methanesulfonylphenyl acetamide, n-2-methylsulfonyl phenyl acetamide, n-2-methylsulfonylphenyl acetamide, n-acetyl-2-methylsulfonyl aniline, n-acetyl-2-methylsulphonyl aniline
PubChem CID 826278
IUPAC Name N-(2-methylsulfonylphenyl)acetamide
SMILES CC(=O)NC1=CC=CC=C1S(=O)(=O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2690703
View Documents
Alfa Aesar™
H2690703
1g
Each for $28.82
 
AAH2690706
View Documents
Alfa Aesar™
H2690706
5g
Each for $101.88
 
AAH2690714
View Documents
Alfa Aesar™
H2690714
25g
Each for $335.07
 
Description

Description

Specifications

Chemical Identifiers

20628-27-7
213.251
NGWCPCSZLLJKPD-UHFFFAOYSA-N
826278
CC(=O)NC1=CC=CC=C1S(=O)(=O)C
C9H11NO3S
MFCD01464107
2'-methylsulfonyl acetanilide, 2'-methylsulfonylacetanilide, acmc-1ceiy, n-2-methanesulfonyl phenyl acetamide, n-2-methanesulfonylphenyl acetamide, n-2-methylsulfonyl phenyl acetamide, n-2-methylsulfonylphenyl acetamide, n-acetyl-2-methylsulfonyl aniline, n-acetyl-2-methylsulphonyl aniline
N-(2-methylsulfonylphenyl)acetamide

Specifications

146°C to 148°C
20628-27-7
MFCD01464107
NGWCPCSZLLJKPD-UHFFFAOYSA-N
N-(2-methylsulfonylphenyl)acetamide
826278
96%
1g
C9H11NO3S
2'-methylsulfonyl acetanilide, 2'-methylsulfonylacetanilide, acmc-1ceiy, n-2-methanesulfonyl phenyl acetamide, n-2-methanesulfonylphenyl acetamide, n-2-methylsulfonyl phenyl acetamide, n-2-methylsulfonylphenyl acetamide, n-acetyl-2-methylsulfonyl aniline, n-acetyl-2-methylsulphonyl aniline
CC(=O)NC1=CC=CC=C1S(=O)(=O)C
213.251
213.26
2'-(Methylsulfonyl)acetanilide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Product Certifications